Learn More
Gold Biotechnology Inc 1 kb DNA Ladder 500 µL
SDP
Catalog No. 501532759
Encompass_Preferred
A unique combination of a number of proprietary plasmids digested with appropriate restriction enzymes and PCR products to yield 13 fragments, suitable for use as molecular weight standards for agarose gel electrophoresis. The DNA includes fragments ranging from 250-10,000 base pairs. The 1K and 3K bands have increased intensity to serve as reference points. The approximate mass of DNA in each band is provided (0.5 μg a load) for approximating the mass of DNA in comparably intense samples. This ladder is ready-to-use, and premixed with loading buffer dye for direct loading on gel.
Catalog No. 50-153-2759 Supplier Gold Biotechnology Inc Supplier No. D010500
A unique combination of a number of proprietary plasmids digested with appropriate restriction enzymes and PCR products to yield 13 fragments, suitable for use as molecular weight standards for agarose gel electrophoresis. The DNA includes fragments ranging from 250-10,000 base pairs. The 1K and 3K bands have increased intensity to serve as reference points. The approximate mass of DNA in each band is provided (0.5 μg a load) for approximating the mass of DNA in comparably intense samples. This ladder is ready-to-use, and premixed with loading buffer dye for direct loading on gel.