Learn More
Ubpbio 1 Kb DNA Ladder, 6X500UL
Catalog No. 501965071
Encompass_Preferred
A unique combination of a number of proprietary plasmids digested with appropriate restriction enzymes and PCR products to yield 13 fragments, suitable for use as molecular weight standards for agarose gel electrophoresis. The DNA includes fragments ranging from 250-10,000 base pairs. The 1K and 3K bands have increased intensity to serve as reference points.The approximate amount of DNA in each band is provided (0.5 μg per load).
Catalog No. 50-196-5071 Supplier Ubpbio Supplier No. L1021
A unique combination of a number of proprietary plasmids digested with appropriate restriction enzymes and PCR products to yield 13 fragments, suitable for use as molecular weight standards for agarose gel electrophoresis. The DNA includes fragments ranging from 250-10,000 base pairs. The 1K and 3K bands have increased intensity to serve as reference points.The approximate amount of DNA in each band is provided (0.5 μg per load).