Learn More
Sigma Aldrich Fine Chemicals Biosciences 1 kb DNA Ladder for DNA el
Catalog No. 502901364
Encompass
Sigmas 1 kb Ladder contains 11 fragments consisting of 500 bp repeats from 0.5 to 3 kb 1 kb repeats from 3 to 6 kb and 2 kb repeats from 6 to 10 kb. In nucleic acid gel electrophoresis five mul of the marker should be diluted in gel loading buffer and then loaded in a single lane on an agarose or polyacrylamide gel. Suitable for use in Northern and Southern blotting. All DNA markers can be stained with ethidium bromide SYBR(R) Green and Nancy-520.
Catalog No. 50-290-1364 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D04281VL
Sigmas 1 kb Ladder contains 11 fragments consisting of 500 bp repeats from 0.5 to 3 kb 1 kb repeats from 3 to 6 kb and 2 kb repeats from 6 to 10 kb. In nucleic acid gel electrophoresis five mul of the marker should be diluted in gel loading buffer and then loaded in a single lane on an agarose or polyacrylamide gel. Suitable for use in Northern and Southern blotting. All DNA markers can be stained with ethidium bromide SYBR(R) Green and Nancy-520.