Learn More
Genscript Corporation 1 kb DNA Marker
Catalog No. 5000058661
Encompass
1 kb DNA Marker is composed of 9 bands which are 500 bp 1 000 bp 2 000 bp 3 000 bp 4 000 bp 5 000 bp 6 000 bp 8 000 bp and 10 000 bp The 5 000 bp band with higher concentration is distinguished easily from others
Catalog No. 50-000-58661 Supplier Genscript Corporation Supplier No. M009915
1 kb DNA Marker is composed of 9 bands which are 500 bp 1 000 bp 2 000 bp 3 000 bp 4 000 bp 5 000 bp 6 000 bp 8 000 bp and 10 000 bp The 5 000 bp band with higher concentration is distinguished easily from others