Learn More
Zymo Research Corporation ZR 1 kb DNA Marker™ (50 μg / 100 μl)
SDP
Catalog No. 501251646
Encompass_Preferred
The 1 kb DNA marker consists of plasmid DNA that has been digested with endonucleases to yield 10 defined fragments that range in size from 0.5 - 10 kb. The intensity of the 3 kb band is increased as an identifier. All fragments have 4-base, 5 overhangs.
Catalog No. 50-125-1646 Supplier Zymo Research Corporation Supplier No. M500350
The 1 kb DNA marker consists of plasmid DNA that has been digested with endonucleases to yield 10 defined fragments that range in size from 0.5 - 10 kb. The intensity of the 3 kb band is increased as an identifier. All fragments have 4-base, 5 overhangs.