Learn More
Gold Biotechnology Inc 1 kb PLUS™ DNA Ladder 500 µL
SDP
Catalog No. 501532760
Encompass_Preferred
The 1 kb PLUSTM is composed of 14 individual DNA fragments: 25K, 10K, 8K, 6K, 5K, 4K, 3K, 2.5K, 2K, 1500, 1000, 750, 500, 250 base pairs. This product contains two enhanced bands (3K and 1K bp) for easy reference. The ladder is ready-to-use, and is premixed with loading buffer dye for direct loading on gel.
Catalog No. 50-153-2760 Supplier Gold Biotechnology Inc Supplier No. D011500
The 1 kb PLUSTM is composed of 14 individual DNA fragments: 25K, 10K, 8K, 6K, 5K, 4K, 3K, 2.5K, 2K, 1500, 1000, 750, 500, 250 base pairs. This product contains two enhanced bands (3K and 1K bp) for easy reference. The ladder is ready-to-use, and is premixed with loading buffer dye for direct loading on gel.