Learn More
Cayman Chemical 1-Kestose, Purity: ≥95%, CAS: 470-69-9, 250 mg
SDP
Catalog No. 502949942
Encompass_Preferred
A fructooligosaccharide with prebiotic and antidiabetic activities; increases levels of the beneficial bacterium B. longum in diluted adult human fecal samples in culture broth at 0.5% w/v; dietary supplementation increases the proportion of cecal Anaerostipes and Bifidobacteriumcontent, improves glucose tolerance, and prevents the development of type 2 diabetes in OLETF rats
Catalog No. 50-294-9942 Supplier Cayman Chemical Supplier No. 34346250 MG
A fructooligosaccharide with prebiotic and antidiabetic activities; increases levels of the beneficial bacterium B. longum in diluted adult human fecal samples in culture broth at 0.5% w/v; dietary supplementation increases the proportion of cecal Anaerostipes and Bifidobacteriumcontent, improves glucose tolerance, and prevents the development of type 2 diabetes in OLETF rats