Learn More
Charter Medical 1 L CELL EXPANSION BAGS 12/CS
Catalog No. NC1856246
Encompass
380mL fill volume at 1cm thick cell expansion bag with two leadsand female luers, and docakble fill line with sampling valve
This item is not returnable. View return policy
Catalog No. NC1856246 Supplier Charter Medical Supplier No. EXP1L
380mL fill volume at 1cm thick cell expansion bag with two leadsand female luers, and docakble fill line with sampling valve