Learn More
MSC Devcon 10110 1 Lb Pail Two Part Epoxy 45 min Working Time, Series Plastic Steel
Catalog No. 501113842
Encompass
Devcon 10110 1 Lb Pail Two Part Epoxy 45 min Working Time, Series Plastic Steel
Catalog No. 50-111-3842 Supplier MSC Supplier No. 00260059
Devcon 10110 1 Lb Pail Two Part Epoxy 45 min Working Time, Series Plastic Steel