Learn More
Medchemexpress LLC 1-Linoleoyl glycerol | 2277-28-3 | 99.52% | 354.52 | 50 MG
SDP
Catalog No. 5000343139
Encompass_Preferred
1-Linoleoyl glycerol is an LpPLA2 inhibitor that acts as a precursor for synthesizing various functional lipids, such as phospholipids. It also mitigates inflammation induced by Apolipoprotein CIII (reduction of IL-6).
  • Acts as an LpPLA2 inhibitor.
  • Precursor for synthesizing various functional lipids.
  • Mitigates inflammation induced by Apolipoprotein CIII.
  • (R)-1-Linoleoyl Glycerol and (S)-1-Linoleoyl Glycerol exhibit Lp-PLA2 inhibitory activities.
  • Metabolized by human eosinophils to 13-HODE-G and 13-HODE.
  • Metabolized by human neutrophils to produce 15-LO metabolite.
Catalog No. 50-003-43139 Supplier Medchemexpress LLC Supplier No. HY111346500MG
1-Linoleoyl glycerol is an LpPLA2 inhibitor that acts as a precursor for synthesizing various functional lipids, such as phospholipids. It also mitigates inflammation induced by Apolipoprotein CIII (reduction of IL-6).
  • Acts as an LpPLA2 inhibitor.
  • Precursor for synthesizing various functional lipids.
  • Mitigates inflammation induced by Apolipoprotein CIII.
  • (R)-1-Linoleoyl Glycerol and (S)-1-Linoleoyl Glycerol exhibit Lp-PLA2 inhibitory activities.
  • Metabolized by human eosinophils to 13-HODE-G and 13-HODE.
  • Metabolized by human neutrophils to produce 15-LO metabolite.