Learn More
Integrated DNA Technologies Inc IDTE pH 7.5 (1X TE Solution), 1L
Catalog No. NC1631631
Encompass
For initial resuspension and storage of DNA oligos.
Catalog No. NC1631631 Supplier Integrated DNA Technologies Inc Supplier No. 11050105
This product needs to be quoted. Please contact your Fisher Scientific Sales Representative for pricing.
For initial resuspension and storage of DNA oligos.