Learn More
Glas Col LLC Heating Mantle
SDP
Catalog No. NC0579334
Encompass_Preferred
Hemispherical fabric heating mantle for 1L single or multi neck flasks. To 400 degrees C maimum internal operating temperature, 380W, 115V, CSA.
Catalog No. NC0579334 Supplier Glas Col LLC Supplier No. 100A O408
Hemispherical fabric heating mantle for 1L single or multi neck flasks. To 400 degrees C maimum internal operating temperature, 380W, 115V, CSA.