Learn More
GFS Chemicals Inc WATERMARK KARL FISCHER COULOMETRIC VESSEL SOLUTION, PYRIDINE-FREE | 1 L | GFS CHEMICALS, INC. Item #: 99771 | Bulk #: 1612
SDP
Catalog No. NC9729157
Encompass_Preferred
WATERMARK KARL FISCHER COULOMETRIC VESSEL SOLUTION, PYRIDINE-FREE | Anolyte for coulometric titration for cells with and without diaphragm
Catalog No. NC9729157 Supplier GFS Chemicals Inc Supplier No. 99771
WATERMARK KARL FISCHER COULOMETRIC VESSEL SOLUTION, PYRIDINE-FREE | Anolyte for coulometric titration for cells with and without diaphragm