Learn More
Chemglass Life Sciences 1000mL Addition Funnel, Graduated, 24/40 Joint Size, 0-4mm PTFE Valve, 550mm OAH
SDP
Catalog No. CG171406
Encompass_Preferred
1000mL Addition Funnel, Graduated, 24/40 Joint Size, 0-4mm PTFE Valve, 550mm OAH. Cylindrical style graduated addition funnel has a threaded, grease-free extended tip valve. Design of all PTFE plug permits semi-needle valve action. With a standard taper top outer joint and lower inner joint, both being the same size, and pressure equalizing arm. Funnels with 14/20 and 19/22 joints are supplied complete with a stopper. For replacement stoppers see CG-3000.
Catalog No. CG171406 Supplier Chemglass Life Sciences Supplier No. CG171406
1000mL Addition Funnel, Graduated, 24/40 Joint Size, 0-4mm PTFE Valve, 550mm OAH. Cylindrical style graduated addition funnel has a threaded, grease-free extended tip valve. Design of all PTFE plug permits semi-needle valve action. With a standard taper top outer joint and lower inner joint, both being the same size, and pressure equalizing arm. Funnels with 14/20 and 19/22 joints are supplied complete with a stopper. For replacement stoppers see CG-3000.