Learn More
Cole-Parmer Blue Enameled Steel and Glass Dewar Flask, 1.1 L
Supplier:  Cole-Parmer 0377434
Encompass_Preferred
The flasks safely transport liquid gases. They have a tough, blue stove-enameled steel outer container and are rust and corrosion resistant. The inside is ultra-low evacuated, high-quality borosilicate glass. They are ideal for use with cold traps. Its cushioned base prevents benchtop damage.
Catalog No. NC1259001