Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1 M 9 9-dimethylfluorene In Thf A Precursor Solution To Prepare The Prelithiation Reagent For Si Anode

Catalog No. 9180595ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-805-95ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 91-805-95ML Supplier Sigma Aldrich Supplier No. 9180595ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

This product offered here allows an ambient-air-tolerant method for negative electrode prelithiation prior to assembly of Li ion batteries. The redox potential of this product makes it useful for those most reported negative electrodes including Si with high capacities and short processing time. It could not only compensate for the initial active lithium loss, but also enable the pairing with high-capacity lithium-free positive electrodes (e.g. sulfur, oxygen, V2O5 etc.) that requires the supply of Li source from the negative electrode to configure Li ion batteries with high energy densities.Recommended instructions (this should be a separate document):Soak your electrode with the anode material coated on a copper current collector in this prelithiation reagent for a set amount of time. Discard the solution and then wash your electrode twice with THF. For a 1cm diameter electrode with 1GO+3 mg acitve material per cm2, use 0.5GO+1 mL of prelithiation solution will be enough. You can use an empty coin cell casing as the treatment vessel. Electrode soaking time is important and may need to be optimized for your application. A good time to try is GO+10 min. Longer prelithiation time may cause eletrode structure deformation, leading to a decreased discharge/charge capacity.

Specifications

CAS 4569-45-3
Molecular Formula C15H14
Linear Formula C15H14
Quantity 5 mL
MDL Number MFCD00114670
Molecular Weight (g/mol) 194.27
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.