Learn More
BIOCLONE INC 1 µm BcMag™ Cleavable Azide-Activated Magnetic Beads – 300 mg
Catalog No. NC3931038
Encompass_Preferred
Efficiently enrich alkyne-tagged biomolecules using BcMag™ Cleavable Azide-Activated Magnetic Beads via Cu(I)-catalyzed Alkyne-Azide (CUAAC) reaction. Benefit from high stability, low nonspecific binding, and superior handling for genomics, proteomics, biomarker discovery, and PTM analysis.
Catalog No. NC3931038 Supplier BIOCLONE INC Supplier No. IJ102
Efficiently enrich alkyne-tagged biomolecules using BcMag™ Cleavable Azide-Activated Magnetic Beads via Cu(I)-catalyzed Alkyne-Azide (CUAAC) reaction. Benefit from high stability, low nonspecific binding, and superior handling for genomics, proteomics, biomarker discovery, and PTM analysis.