Learn More
SPEX CERTIPREP LLC 3P 1% MAGNESIUM NITRATE (MG 250ML
Catalog No. NC1811365
Encompass_Preferred
(HPS by Zeptometrix) 1% Magnesium Nitrate (MgNO3) Matrix Modifier, 250mL
Catalog No. NC1811365 Supplier SPEX CERTIPREP LLC 3P Supplier No. MM9011250
(HPS by Zeptometrix) 1% Magnesium Nitrate (MgNO3) Matrix Modifier, 250mL