Learn More
Uline 1"" MASKING TAPE, PACK OF 12
Catalog No. NC1466212
Encompass
Tough industrial tape for paint masking and bundling.Adheres to stainless steel, powder coatings, EPDM rubber and alodine aluminum.Good paint line. Clean removal. Resists solvents and moisture.Withstands 250¿F for up to 30 minutes.Smooth tan crepe paper backing with rubber adhesive
Catalog No. NC1466212 Supplier Uline Supplier No. S5698PK
Tough industrial tape for paint masking and bundling.Adheres to stainless steel, powder coatings, EPDM rubber and alodine aluminum.Good paint line. Clean removal. Resists solvents and moisture.Withstands 250¿F for up to 30 minutes.Smooth tan crepe paper backing with rubber adhesive