Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Mercapto-2-propanol

Catalog No. 3283755G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-837-55G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 32-837-55G Supplier Sigma Aldrich Supplier No. 3283755G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Mercapto-2-propanol is a chain transferring agent that is the sixth series of hydrogen bonded 1,2-propanols which can be used to form polymeric chains with low molecular weight and short chain length.1-Mercapto-2-propanol can be used as a self-assembled monolayer on the gold surfaces with potential use in electrochemical applications. It may also be used as a chiral agent which can be used in the chemical analysis of enantiomers.

Specifications

CAS 1068-47-9
Density 1.048 g/mL (at 25°C)
Boiling Point 58°C to 60°C (7 mmHg)
Molecular Formula C3H8OS
Linear Formula CH3CH(OH)CH2SH
Refractive Index n20/D 1.486 (literature)
Quantity 5 g
MDL Number MFCD00004878
Molecular Weight (g/mol) 92.16
Percent Purity 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.