Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(Mercaptomethyl)cyclopropaneacetic Acid 97.0+%, TCI America™
SDP

Catalog No. M20745G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20745G 5 g
M20741G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M20745G Supplier TCI America Supplier No. M20745G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 162515-68-6
Molecular Formula C6H10O2S
Molecular Weight (g/mol) 146.204
MDL Number MFCD03411600
InChI Key VFAXPOVKNPTBTM-UHFFFAOYSA-N
Synonym 2-1-mercaptomethyl cyclopropyl acetic acid, 1-mercaptomethyl cyclopropaneacetic acid, 1-mercaptomethyl cyclopropylacetic acid, 1-mercaptomethyl-cyclopropaneacetic acid, unii-1x00t93wyx, cyclopropaneacetic acid, 1-mercaptomethyl, 2-1-sulfanylmethyl cyclopropyl acetic acid, 1-mercaptomethyl cyclopropyl acetic acid, 1-sulfanylmethyl cyclopropyl acetic acid
PubChem CID 9793825
IUPAC Name 2-[1-(sulfanylmethyl)cyclopropyl]acetic acid
SMILES C1CC1(CC(=O)O)CS

Specifications

Melting Point 45°C
Color White-Yellow
Quantity 5 g
UN Number 3261
Formula Weight 146.20
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(Mercaptomethyl)cyclopropaneacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.