Learn More
Metrohm USA 1 METER CABLE G TO F PLUG
SDP
Supplier:  Metrohm USA 62104020
Encompass
1 meter cable for Double Platinum Electrode for connecting electrodes with Metrohm plug-in head G to Metrohm instruments (socket F). For use with Karl Fischer volumetric titrator.
Catalog No. NC9816796