Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methoxy-2-indanol 98%, Thermo Scientific™

Catalog No. AC186100250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC186100250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC186100250 Supplier Thermo Scientific Chemicals Supplier No. 186100250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56175-44-1
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
InChI Key ZHWPZRVYPHGXSF-UHFFFAOYSA-N
Synonym 1-methoxy-2-indanol, 1h-inden-2-ol, 2,3-dihydro-1-methoxy-, cis, 1h-inden-2-ol,2,3-dihydro-1-methoxy-, 1r,2s-rel, 1-methoxyindan-2-ol, 1-methoxy-2-indanol #, zhwpzrvyphgxsf-uhfffaoysa, 1h-inden-2-ol, 2,3-dihydro-1-methoxy
PubChem CID 143339
IUPAC Name 1-methoxy-2,3-dihydro-1H-inden-2-ol
SMILES COC1C(CC2=CC=CC=C12)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Brown
Infrared Spectrum Authentic
Refractive Index 1.5475 to 1.5495 (20°C, 589nm)
Quantity 25 g
Beilstein 06, 970
Formula Weight 164.2
Percent Purity ≥97.5%
Physical Form Viscous Liquid
Chemical Name or Material 1-Methoxy-2-indanol, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.