Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methoxy-2-methyl-2-propanol, 98%, AcroSeal™, Thermo Scientific™

Catalog No. AC381711000
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC381711000 100 mL AcroSeal glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC381711000 Supplier Thermo Scientific Chemicals Supplier No. 381711000

missing translation for 'validMainframeMsg'

 

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3587-64-2
Molecular Formula C5H12O2
Molecular Weight (g/mol) 104.15
MDL Number MFCD03701584
InChI Key MXUXZWFVAPTPAG-UHFFFAOYSA-N
Synonym 1-methoxy-2-methyl-2-propanol, 2-propanol, 1-methoxy-2-methyl, acmc-20aott, 2-methoxymethyl-2-propanol, 1,1-dimethyl-2-methoxyethanol, 2-propanol,1-methoxy-2-methyl, 1-methoxy-2-methyl-2-propanol #
PubChem CID 77137
IUPAC Name 1-methoxy-2-methylpropan-2-ol
SMILES COCC(C)(C)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 0.8970g/mL
Boiling Point 108.0°C to 110.0°C
Flash Point 28°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging AcroSeal glass Bottle
Refractive Index 1.4025 to 1.4065 (20°C, 589nm)
Linear Formula CH3OCH2C(CH3)2OH
Quantity 100 mL
Specific Gravity 0.897
Solubility Information Solubility in water: miscible
Formula Weight 104.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Methoxy-2-methyl-2-propanol
Show More Show Less
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.