Learn More
1-Methoxy-5-Methylphenazinium Methyl Sulfate, Sigma-Aldrich
Shop All MilliporeSigma Supelco ProductsDescription
1-Methoxy-5-methylphenazinium methyl sulfate (MPMS) is a photochemically stable electron-transport mediator between NAD(P)H and tetrazolium dyes. The reduced form of MPMS converts tetrazolium salt to purple precipitate formazan. It is a very stable and non-enzymic electron carrier that mediates electron transfer by dehydrogenases in vitro to various electron acceptors such as tetrazolium dyes or the electrode of an enzymic electric cell.
Specifications
Specifications
| Detection Method | Fluorescence |
| Concentration | ≥95% |
| Format | Bottle |
| Formula Weight | 336.36 g/mol |
| Product Type | Stain |
| Quantity | 500 mg |
| Form | Powder |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.