Learn More
Selleck Chemical LLC 1-Methoxy PMS
SDP
Catalog No. 5000001065
Encompass_Preferred
Methoxy-PMS (1-Methoxy PMS) an active oxygen formation inducer is a stable electron-transport mediator between NAD(P)H and tetrazolium dyes Methoxy-PMS receives an electron from NADH or NADPH at the membrane or inside of the cell and passes the electron to the WST-8 that is around the outer cell membrane
Catalog No. 50-000-01065 Supplier Selleck Chemical LLC Supplier No. S7770100MG
Methoxy-PMS (1-Methoxy PMS) an active oxygen formation inducer is a stable electron-transport mediator between NAD(P)H and tetrazolium dyes Methoxy-PMS receives an electron from NADH or NADPH at the membrane or inside of the cell and passes the electron to the WST-8 that is around the outer cell membrane