Learn More
Selleck Chemical LLC 1-Methoxy PMS-25mg
SDP
Supplier:  Selleck Chemical LLC S777025MG
Encompass_Preferred
Methoxy-PMS (1-Methoxy PMS), an active oxygen formation inducer, is a stable electron-transport mediator between NAD(P)H and tetrazolium dyes. Methoxy-PMS receives an electron from NADH or NADPH at the membrane or inside of the cell and passes the electron to the WST-8 that is around the outer cell membrane.
Catalog No. 50-238-9941