Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-Methoxycarbonyl-β-carboline | 3464-66-2 | 226.23 | 5 MG
SDP

Catalog No. 5000416001
Encompass_Preferred

1-Methoxycarbonyl-β-carboline is a natural alkaloid intended for research use only. This solid, white to off-white compound has a purity of 98.77%.

  • Natural alkaloid
  • High purity (98.77%)
  • Solid, white to off-white appearance
  • Classified as pyridine and indole alkaloid
  • Derived from plants (Simaroubaceae)
  • Soluble in DMSO (100 mg/mL)
  • Recommended storage at 4°C, protected from light (solid form)
  • Recommended storage at -80°C for 6 months or -20°C for 1 month (in solvent)

Catalog No. 50-004-16001 Supplier Medchemexpress LLC Supplier No. HYN16335MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

1-Methoxycarbonyl-β-carboline is a natural alkaloid intended for research use only. This solid, white to off-white compound has a purity of 98.77%.

  • Natural alkaloid
  • High purity (98.77%)
  • Solid, white to off-white appearance
  • Classified as pyridine and indole alkaloid
  • Derived from plants (Simaroubaceae)
  • Soluble in DMSO (100 mg/mL)
  • Recommended storage at 4°C, protected from light (solid form)
  • Recommended storage at -80°C for 6 months or -20°C for 1 month (in solvent)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.