Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-Methoxycyclopropanecarboxylic acid | 100683-08-7 | MFCD19228478 | ≥98.0% | 116.12 g·mol⁻¹ | C5H8O3 | 250 MG
SDP

Catalog No. 5000209189
Encompass_Preferred

1-Methoxycyclopropanecarboxylic acid is a small carboxylic acid used as a mitochondrial uncoupler in biochemical research. It stimulates cellular respiration to about 160% of baseline levels and is supplied for research use only.

  • Purity ≥98.0%.
  • Molecular weight 116.12 g·mol⁻¹.
  • Chemical formula C5H8O3.
  • Available in laboratory pack sizes such as 100 MG, 250 MG, 500 MG, 1 G.
  • Useful as a biochemical reagent for studies of mitochondrial respiration and metabolic regulation.

Catalog No. 50-002-09189 Supplier Medchemexpress LLC Supplier No. 250MGHYW046704
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Methoxycyclopropanecarboxylic acid is a small carboxylic acid used as a mitochondrial uncoupler in biochemical research. It stimulates cellular respiration to about 160% of baseline levels and is supplied for research use only.

  • Purity ≥98.0%.
  • Molecular weight 116.12 g·mol⁻¹.
  • Chemical formula C5H8O3.
  • Available in laboratory pack sizes such as 100 MG, 250 MG, 500 MG, 1 G.
  • Useful as a biochemical reagent for studies of mitochondrial respiration and metabolic regulation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.