Learn More
Medchemexpress LLC 1-Methoxynaphthalene | 2216-69-5 | 98.6% | 158.20 | 100 G
SDP
Catalog No. 5000453029
Encompass_Preferred
1-Methoxynaphthalene is utilized as a substrate to investigate the activity of cytochrome c peroxidase (CcP). It can also be employed in the synthesis of prenyl naphthalen-ols. This product is intended for research use only and is not sold to patients.
  • Used as a substrate for cytochrome c peroxidase (CcP) activity investigations.
  • Can be used to synthesize prenyl naphthalen-ols.
Catalog No. 50-004-53029 Supplier Medchemexpress LLC Supplier No. HYW012568100G
1-Methoxynaphthalene is utilized as a substrate to investigate the activity of cytochrome c peroxidase (CcP). It can also be employed in the synthesis of prenyl naphthalen-ols. This product is intended for research use only and is not sold to patients.
  • Used as a substrate for cytochrome c peroxidase (CcP) activity investigations.
  • Can be used to synthesize prenyl naphthalen-ols.