Learn More
Medchemexpress LLC 1-Methoxynaphthalene | 2216-69-5 | 98.6% | 158.20 | 250 G
SDP
Catalog No. 5000451518
Encompass_Preferred
1-Methoxynaphthalene is utilized as a substrate to explore the activity of cytochrome c peroxidase (CcP). It can also be employed in the synthesis of prenyl naphthalen-ols.
  • Used as a substrate to investigate the activity of cytochrome c peroxidase (CcP).
  • Can be used to synthesize prenyl naphthalen-ols.
Catalog No. 50-004-51518 Supplier Medchemexpress LLC Supplier No. 250GHYW012568
1-Methoxynaphthalene is utilized as a substrate to explore the activity of cytochrome c peroxidase (CcP). It can also be employed in the synthesis of prenyl naphthalen-ols.
  • Used as a substrate to investigate the activity of cytochrome c peroxidase (CcP).
  • Can be used to synthesize prenyl naphthalen-ols.