Learn More
Medchemexpress LLC HY-W012568 500mg Medchemexpress, 1-Methoxynaphthalene CAS:2216-69-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW012568500MG
Encompass_Preferred
Medchemexpress, HY-W012568 500mg 1-Methoxynaphthalene CAS:2216-69-5 1-Methoxynaphthalene is used as the substrate to investigate the activity of cytochrome c peroxidase (CcP). 1-Methoxynaphthalene also can be used to synthesize prenyl naphthalen-ols. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3329