Learn More
Medchemexpress LLC 1-methyl-1,4-dihydronicotinamide | 17750-23-1 | AKOS006277282 | 95.0% | 138.17 g/mol | C7H10N2O | 10MG
SDP
Catalog No. 5000238479
Encompass_Preferred
1-Methyl-1,4-dihydronicotinamide is a reduced nicotinamide derivative (CAS 17750-23-1) used as a research reagent with reported radical-scavenging activity. It is supplied as a yellow to orange solid, molecular formula C7H10N2O, molecular weight 138.17 g/mol, and typical purity of 95.0%.
  • Radical scavenging activity reported in biochemical studies.
  • Provided as a solid for preparation of laboratory stock solutions.
  • Molecular formula C7H10N2O and molecular weight 138.17 g/mol.
  • Typical purity 95.0% appropriate for research applications.
  • Available in small mg-scale pack sizes for experimental use.
Catalog No. 50-002-38479 Supplier Medchemexpress LLC Supplier No. HY12431310MG
1-Methyl-1,4-dihydronicotinamide is a reduced nicotinamide derivative (CAS 17750-23-1) used as a research reagent with reported radical-scavenging activity. It is supplied as a yellow to orange solid, molecular formula C7H10N2O, molecular weight 138.17 g/mol, and typical purity of 95.0%.
  • Radical scavenging activity reported in biochemical studies.
  • Provided as a solid for preparation of laboratory stock solutions.
  • Molecular formula C7H10N2O and molecular weight 138.17 g/mol.
  • Typical purity 95.0% appropriate for research applications.
  • Available in small mg-scale pack sizes for experimental use.