Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-1-[4-(diphenylphosphino)benzyl]pyrrolidinium Bromide 98.0+%, TCI America™
SDP

Catalog No. M21031G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M21031G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M21031G Supplier TCI America Supplier No. M21031G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1229444-44-3
Molecular Formula C24H27BrNP
Molecular Weight (g/mol) 440.37
MDL Number MFCD16295188
InChI Key JKJSVXHLMBCNPA-UHFFFAOYSA-M
PubChem CID 53384383
IUPAC Name 1-{[4-(diphenylphosphanyl)phenyl]methyl}-1-methylpyrrolidin-1-ium bromide
SMILES [Br-].C[N+]1(CC2=CC=C(C=C2)P(C2=CC=CC=C2)C2=CC=CC=C2)CCCC1

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 440.36
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Methyl-1-[4-(diphenylphosphino)benzyl]pyrrolidinium Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.