Learn More
Medchemexpress LLC 1-methyl-1,5-dihydro-4H-pyrazolo[3,4-d]pyrimidin-4-one | 5334-56-5 | MFCD01312343 | 150.14 g/mol | C6H6N4O | 5 G
SDP
Catalog No. 5000209958
Encompass_Preferred
1-methyl-1,5-dihydro-4H-pyrazolo[3,4-d]pyrimidin-4-one (CAS 5334-56-5) is a pyrazolopyrimidinone small-molecule used as a biochemical reagent and reference compound in life-science research.
  • Chemical formula C6H6N4O, molecular weight 150.14 g/mol.
  • Solid form with a reported melting point >250 °C.
  • Commonly used as a biochemical reagent and reference compound in pharmacological and biochemical studies.
  • Typical supplier package sizes include small gram quantities for research use.
  • Reported purities vary by supplier; verify the exact purity from the product-specific safety data sheet or manufacturer listing.
Catalog No. 50-002-09958 Supplier Medchemexpress LLC Supplier No. HYW0522495G
1-methyl-1,5-dihydro-4H-pyrazolo[3,4-d]pyrimidin-4-one (CAS 5334-56-5) is a pyrazolopyrimidinone small-molecule used as a biochemical reagent and reference compound in life-science research.
  • Chemical formula C6H6N4O, molecular weight 150.14 g/mol.
  • Solid form with a reported melting point >250 °C.
  • Commonly used as a biochemical reagent and reference compound in pharmacological and biochemical studies.
  • Typical supplier package sizes include small gram quantities for research use.
  • Reported purities vary by supplier; verify the exact purity from the product-specific safety data sheet or manufacturer listing.