Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-methyl-1,5-dihydro-4H-pyrazolo[3,4-d]pyrimidin-4-one | 5334-56-5 | MFCD01312343 | >98.0% | 150.14 g/mol | C6H6N4O | 1 G
SDP

Catalog No. 5000210922
Encompass_Preferred

1-Methyl-1,5-dihydro-4H-pyrazolo[3,4-d]pyrimidin-4-one is a small heterocyclic organic compound used as a biochemical reagent in life-science research. It is supplied as a powder and is used as a reagent or intermediate in chemical synthesis and biochemical assays.

  • Used as a reagent in biochemical assays and synthesis.
  • Small heterocyclic structure suitable for medicinal chemistry applications.
  • CAS number 5334-56-5 for unambiguous identification.
  • Molecular formula C6H6N4O and molecular weight ~150.14 g/mol.
  • Typical physical form: powder; common pack sizes include 1 g and 5 g.
  • Refer to lot-specific certificate of analysis for purity and storage conditions.

Catalog No. 50-002-10922 Supplier Medchemexpress LLC Supplier No. HYW0522491G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Methyl-1,5-dihydro-4H-pyrazolo[3,4-d]pyrimidin-4-one is a small heterocyclic organic compound used as a biochemical reagent in life-science research. It is supplied as a powder and is used as a reagent or intermediate in chemical synthesis and biochemical assays.

  • Used as a reagent in biochemical assays and synthesis.
  • Small heterocyclic structure suitable for medicinal chemistry applications.
  • CAS number 5334-56-5 for unambiguous identification.
  • Molecular formula C6H6N4O and molecular weight ~150.14 g/mol.
  • Typical physical form: powder; common pack sizes include 1 g and 5 g.
  • Refer to lot-specific certificate of analysis for purity and storage conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.