Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-1-cyclohexene, 97%, stabilized

Catalog No. AC156841000
Change view
Click to view available options
Quantity:
10 g
25 mL
50 g
100 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC156841000 100 mL
AC156840100 10 g
AC156840500 50 g
AC156840250 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC156841000 Supplier Thermo Scientific Chemicals Supplier No. 156841000

CAS: 591-49-1 | C7H12 | 96.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 591-49-1
InChI Key CTMHWPIWNRWQEG-UHFFFAOYSA-N
Synonym 1-methyl-1-cyclohexene, cyclohexene, 1-methyl, 1-methylcyclohex-1-ene, cyclohexene, methyl, methylcyclohexene, .alpha.-methylcyclohexene, 1-methyl-cyclohexene, methyl-1-cyclohexene, 2,3,4,5-tetrahydrotoluene, unii-te4p8q2044
PubChem CID 11574
IUPAC Name 1-methylcyclohexene
SMILES CC1=CCCCC1

Specifications

CAS Min % 98.0
CAS Max % 100.0
Color Colorless
Density 0.81g/mL
Infrared Spectrum Authentic
Assay Percent Range 98+%
Molecular Formula C7H12
Refractive Index 1.4492 to 1.4512 (20°C, 589nm)
Linear Formula C6H9CH3
Quantity 100 mL
Beilstein 05, 66
Stabilizer 10ppm hydroquinone
Molecular Weight (g/mol) 96.173
Formula Weight 96.17
Percent Purity ≥98.0%
Physical Form Liquid
Chemical Name or Material 1-Methyl-1-cyclohexene, 98+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 209-718-0
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.