Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-1-(m-tolyl)hydrazine 97.0+%, TCI America™
SDP

Catalog No. M14195G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M14195G 5 g
M141925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M14195G Supplier TCI America Supplier No. M14195G

Chemical Identifiers

CAS 72233-91-1
Molecular Formula C8H12N2
Molecular Weight (g/mol) 136.20
MDL Number MFCD03844802
InChI Key ABANUKHECQXXRH-UHFFFAOYSA-N
PubChem CID 827964
IUPAC Name 1-methyl-1-(3-methylphenyl)hydrazine
SMILES CN(N)C1=CC=CC(C)=C1

Specifications

Quantity 5 g
Formula Weight 136.20
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material 1-Methyl-1-(m-tolyl)hydrazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.