Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-1-propylpiperidinium Bis(fluorosulfonyl)imide 98.0+%, TCI America™
SDP

Catalog No. M29811G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M29811G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M29811G Supplier TCI America Supplier No. M29811G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 911303-46-3
Molecular Formula C9H20F2N2O4S2
Molecular Weight (g/mol) 322.39
MDL Number MFCD22460465
InChI Key BGMNWBQPCOEGHI-UHFFFAOYSA-N
PubChem CID 121234260
IUPAC Name 1-methyl-1-propylpiperidin-1-ium; bis(fluorosulfonyl)azanide
SMILES FS(=O)(=O)[N-]S(F)(=O)=O.CCC[N+]1(C)CCCCC1

Specifications

Color Yellow
Quantity 1 g
Formula Weight 322.39
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 1-Methyl-1-propylpiperidinium Bis(fluorosulfonyl)imide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.