Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1-Methyl-1H-imidazol-4-yl)methylamine, 95%, Thermo Scientific™

Catalog No. CC03613DE
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC03613DE 5 g Amber Glass Bottle
CC03613CB 250 mg Amber Glass Bottle
CC03613DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC03613DE Supplier Thermo Scientific Chemicals Supplier No. CC03613DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 486414-83-9
Molecular Formula C5H9N3
Molecular Weight (g/mol) 111.15
MDL Number MFCD06411536
InChI Key YSEAGFBRAQOCFM-UHFFFAOYSA-N
PubChem CID 2795114
IUPAC Name (1-methylimidazol-4-yl)methanamine
SMILES CN1C=NC(CN)=C1

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
Synonym 1-methyl-1h-imidazol-4-yl methanamine, 1-methyl-1h-imidazol-4-yl methylamine, 1h-imidazole-4-methanamine, 1-methyl, 1-methylimidazol-4-yl methanamine, 4-aminomethyl-1-methylimidazole, c-1-methyl-1h-imidazol-4-yl-methylamine, 1-1-methylimidazol-4-yl methanamine, 4-aminomethyl-1-methyl-1h-imidazole, 1h-imidazole-4-methanamine,1-methyl, 1-methyl-1h-imidazol-4-yl methyl amine
Formula Weight 111.15
Percent Purity 95%
Chemical Name or Material (1-Methyl-1H-imidazol-4-yl)methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.