Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1-Methyl-1H-imidazol-5-yl)methylamine, 97%, Thermo Scientific™

Catalog No. CC23613DE
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC23613DE 5 g Amber Glass Bottle
CC23613CB 250 mg Amber Glass Bottle
CC23613DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC23613DE Supplier Thermo Scientific Chemicals Supplier No. CC23613DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 486414-86-2
Molecular Formula C5H9N3
Molecular Weight (g/mol) 111.148
InChI Key PYAQTQXFMQWCHQ-UHFFFAOYSA-N
Synonym 1-methyl-1h-imidazol-5-yl methylamine, 1-methyl-5-aminomethylimidazole, 1-methyl-1h-imidazol-5-yl methanamine, 3-methylimidazol-4-yl methanamine, 1h-imidazole-5-methanamine, 1-methyl, 5-aminomethyl-1-methyl-1h-imidazole, c-3-methyl-3h-imidazol-4-yl-methylamine, 1-3-methylimidazol-4-yl methanamine, 1-methyl-imidazol-5-yl methylamine, 1-1-methyl-1h-imidazole-5-yl methanamine
PubChem CID 2795424
IUPAC Name (3-methylimidazol-4-yl)methanamine
SMILES CN1C=NC=C1CN

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 111.15
Percent Purity 97%
Chemical Name or Material (1-Methyl-1H-imidazol-5-yl)methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.