Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-1H-imidazole-4-carbaldehyde, 95%, Thermo Scientific™

Catalog No. CC03604DA
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC03604DA 1 g Amber Glass Bottle
CC03604CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC03604DA Supplier Thermo Scientific Chemicals Supplier No. CC03604DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 17289-26-8
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.116
MDL Number MFCD03411957
InChI Key CQZXDIHVSPZIGF-UHFFFAOYSA-N
PubChem CID 573419
IUPAC Name 1-methylimidazole-4-carbaldehyde
SMILES CN1C=C(N=C1)C=O

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 1-methyl-1h-imidazole-4-carbaldehyde, 1-methylimidazole-4-carboxaldehyde, 1-methyl-1h-imidazole-4-carboxaldehyde, 1h-imidazole-4-carboxaldehyde, 1-methyl, methyl-1h-imidazole-4-carbaldehyde, 1-methylimidazol-4-carboxaldehyde, 4-formyl-1-methyl-1h-imidazole, 1-methyl-4-formyl-imidazole, 1-methyl-1h-imidazole-4-carbaldehyde #, 1h-imidazole-4-carboxaldehyde,1-methyl
Formula Weight 110.12
Percent Purity 95%
Chemical Name or Material 1-Methyl-1H-imidazole-4-carbaldehyde
Hazard Category Warning
Hazard Statement GHS P Statement Causes skin irritation. Harmful if swallowed. May cause respiratory irritation. Causes serious eye irritation.
Precautionary Statement GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protec
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.