Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-methyl-1h-indazole-3-carbonyl chloride, 97%, Thermo Scientific™

Catalog No. CC06302EA
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC06302EA 10 g Amber Glass Bottle
CC06302CB 250 mg Amber Glass Bottle
CC06302DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC06302EA Supplier Thermo Scientific Chemicals Supplier No. CC06302EA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 106649-02-9
Molecular Formula C9H7ClN2O
Molecular Weight (g/mol) 194.62
MDL Number MFCD02093096
InChI Key NPNWVMBPSINFBV-UHFFFAOYSA-N
Synonym 1-methyl-1h-indazole-3-carbonyl chloride, 1h-indazole-3-carbonylchloride, 1-methyl, 1-methyl-1h-indazol-3-carbonyl chloride, 1h-indazole-3-carbonyl chloride, 1-methyl, acmc-20mac8, 1-methyl-3-indazolylcarbonyl chloride, 1-methyl-indazole-3-carbonyl chloride, 1-methyl-1h-indazole-3-carboxychloride, 1-methyl-1h-indazole-3-carboxylic acid chloride
PubChem CID 7022055
IUPAC Name 1-methylindazole-3-carbonyl chloride
SMILES CN1N=C(C(Cl)=O)C2=CC=CC=C12

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 194.62
Percent Purity 97%
Chemical Name or Material 1-Methyl-1H-indazole-3-carbonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.