Learn More
Medchemexpress LLC 1-Methyl-1H-indazole-3-carboxylic acid | 50890-83-0 | 176.18 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW007927
Encompass_Preferred
1-Methyl-1H-indazole-3-carboxylic acid is a biochemical reagent used in life science research. This organic compound, appearing as a white to off-white solid, is stable under recommended storage conditions and is for research use only.
  • Functions as a biological material or organic compound
  • Stable under recommended storage conditions
  • Store at room temperature for up to 3 years
  • In solvent, stable at -80°C for 2 years or -20°C for 1 year
  • Ships at room temperature for short durations
  • Keep in tightly sealed container, cool, well-ventilated area
  • Protect from direct sunlight and sources of ignition
Catalog No. 50-004-48138