Learn More
Sigma Aldrich 1-methyl-1H-indole-4-carboxylic acid
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- 3-ethynylaniline is a terminal alkyne that can be prepared by the reduction of 3-ethylnylnitrobenzene.3-ethynylaniline may be used in the synthesis of the following benzoxazine monomers:
- Bis{4-[3-(3-ethynylphenyl)(2h,4hbenzo[3,4-e]1,3-oxazin-6-yloxy)]phenyl}phenylphosphino-1-one
- [3-(3-ethynylphenyl)(2h,4hbenzo[3,4-e]1,3-oxazaperhydroin-6-yl)][3-(3-ethynylphenyl)(2h,4h-benzo[3,4-e]1,3-oxazin-6-yl)]phenylphosphino-1-one
- Bis[3-(3-ethynylphenyl)(2h,4hbenzo[3,4-e]1,3-oxazin-6-yl)oxy]phenylphosphino-1-oneit may be used:
- To synthesize succin(m-ethynyl)dianilide and sebaco(m-ethynyl)dianilide
- As a reagent in the multi-step synthesis of erlotinib hydrochloride
- To prepare 3-(3-ethynylphenyl)-6-methyl-2h, 4h-benzo[e]1,3-oxazine
- To prepare 3-[3-(4-acetylamino-benzyl)-[1,2,4]oxadiazol-5- yl]-n-(3-ethynyl-phenyl)-propionamide
Specifications
Specifications
| Molecular Formula | C10H9NO2 |
| Linear Formula | C10H9NO2 |
| MDL Number | MFCD08435874 |
| Quantity | 5 g |
| Molecular Weight (g/mol) | 175.18 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.