Learn More
Medchemexpress LLC 1-Methyl-1H-pyrazole-3-carboxylic acid | 25016-20-0 | 99.53% | 126.11 | 10 G
SDP
Catalog No. 5000404978
Encompass_Preferred
1-Methyl-1H-pyrazole-3-carboxylic acid is a chemical intermediate. It is utilized in various synthetic processes within chemical research and development.
  • Used in synthesis of compound 91
  • Acts as a SCD1 and SCD5 inhibitor
  • Relevant for research in metabolic disorders
Catalog No. 50-004-04978 Supplier Medchemexpress LLC Supplier No. HY3460310G
1-Methyl-1H-pyrazole-3-carboxylic acid is a chemical intermediate. It is utilized in various synthetic processes within chemical research and development.
  • Used in synthesis of compound 91
  • Acts as a SCD1 and SCD5 inhibitor
  • Relevant for research in metabolic disorders