Learn More
Medchemexpress LLC 1-methyl-1H-pyrazole-4-boronic acid pinacol ester-d3 | 1022151-50-3 | >98.0% | 211.08 g/mol | C10H14D3BN2O2 | 10 G
SDP
Catalog No. 5000280856
Encompass_Preferred
Deuterated 1-methyl-1H-pyrazole-4-boronic acid pinacol ester-d3 is an isotope-labeled reagent used as a synthetic intermediate and analytical standard. It is provided as a high-purity boronic ester (≥98.0%), with molecular formula C10H14D3BN2O2 and molecular weight 211.08 g/mol, and is intended for use in labeling, method development, and cross-coupling chemistry.
  • Isotope-labeled reagent for synthetic chemistry and tracing.
  • High purity ≥98.0% ensures reliable analytical performance.
  • Pinacol boronic ester functionality suitable for cross-coupling reactions.
  • Molecular formula C10H14D3BN2O2 and molecular weight 211.08 g/mol.
  • Available in a 10 G quantity for preparative applications.
Catalog No. 50-002-80856 Supplier Medchemexpress LLC Supplier No. HYW24951410G
Deuterated 1-methyl-1H-pyrazole-4-boronic acid pinacol ester-d3 is an isotope-labeled reagent used as a synthetic intermediate and analytical standard. It is provided as a high-purity boronic ester (≥98.0%), with molecular formula C10H14D3BN2O2 and molecular weight 211.08 g/mol, and is intended for use in labeling, method development, and cross-coupling chemistry.
  • Isotope-labeled reagent for synthetic chemistry and tracing.
  • High purity ≥98.0% ensures reliable analytical performance.
  • Pinacol boronic ester functionality suitable for cross-coupling reactions.
  • Molecular formula C10H14D3BN2O2 and molecular weight 211.08 g/mol.
  • Available in a 10 G quantity for preparative applications.