Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-methyl-1H-pyrazole-4-boronic acid pinacol ester-d3 | 1022151-50-3 | >98.0% | 211.08 g/mol | C10H14D3BN2O2 | 500 MG
SDP

Catalog No. 5000282334
Encompass_Preferred

1-Methyl-1H-pyrazole-4-boronic acid pinacol ester-d3 is a deuterium-labeled boronic ester supplied for research use as an isotope-labeled synthetic building block. It is suitable for labeled synthesis, mechanistic studies, and mass spectrometry applications where deuterium incorporation aids tracing and quantitation.

  • Deuterium-labeled isotopologue (d3)
  • High purity: >98.0%
  • Molecular weight 211.08 g/mol
  • Chemical formula C10H14D3BN2O2
  • Available in a 500 mg pack size

Catalog No. 50-002-82334 Supplier Medchemexpress LLC Supplier No. HYW249514500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Methyl-1H-pyrazole-4-boronic acid pinacol ester-d3 is a deuterium-labeled boronic ester supplied for research use as an isotope-labeled synthetic building block. It is suitable for labeled synthesis, mechanistic studies, and mass spectrometry applications where deuterium incorporation aids tracing and quantitation.

  • Deuterium-labeled isotopologue (d3)
  • High purity: >98.0%
  • Molecular weight 211.08 g/mol
  • Chemical formula C10H14D3BN2O2
  • Available in a 500 mg pack size

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.