Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl 2-nitroterephthalate, 97%, Thermo Scientific™

Catalog No. AC398000050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC398000050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC398000050 Supplier Thermo Scientific Chemicals Supplier No. 398000050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 35092-89-8
Molecular Formula C9H7NO6
Molecular Weight (g/mol) 225.16
MDL Number MFCD00024510
InChI Key MIIADZYPHVTLPR-UHFFFAOYSA-N
Synonym 4-methoxycarbonyl-3-nitrobenzoic acid, 1-methyl 2-nitroterephthalate, methyl hydrogen 2-nitroterephthalate, 3-nitro-4-carbomethoxybenzoic acid, 2-nitroterephthalic acid 1-methyl ester, methyl 2-nitro-4-carboxybenzoate, methyl-2-nitro-4-carboxy-benzoate, 4-carbomethoxy-3-nitro-benzoic acid
PubChem CID 98592
IUPAC Name 4-(methoxycarbonyl)-3-nitrobenzoic acid
SMILES COC(=O)C1=CC=C(C=C1[N+]([O-])=O)C(O)=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 176.0°C to 178.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 225.16
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 1-Methyl 2-nitroterephthalate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 252-360-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.