Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-2-phenylethyl Acetate 95.0+%, TCI America™
SDP

Catalog No. M14955G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M14955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M14955G Supplier TCI America Supplier No. M14955G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2114-33-2
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.231
MDL Number MFCD01672540
InChI Key LOHDWLZDQXQLJA-UHFFFAOYSA-N
Synonym Acetic Acid 1-Methyl-2-phenylethyl Ester, 2-Acetoxy-1-phenylpropane
PubChem CID 16457
IUPAC Name 1-phenylpropan-2-yl acetate
SMILES CC(CC1=CC=CC=C1)OC(=O)C

Specifications

Color Colorless
Quantity 5 g
Formula Weight 178.23
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Methyl-2-phenylethyl Acetate
RTECSNumber AI9316000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.